Assay Details
Gene Symbol: Ctsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022597.2 | NP_072119.2 | 2 - 3 | 255 | 147 | |
GenBank mRNA | BC072490.1 | - | 2 - 3 | 218 | 147 | |
X82396.1 | - | 2 - 3 | 184 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
