Assay Details
Gene Symbol: Gatm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031031.2 | NP_112293.1 | 8 - 9 | 1220 | 151 | |
GenBank mRNA | BC081785.1 | - | 8 - 9 | 1220 | 151 | |
FQ212973.1 | - | 4 - 5 | 391 | 151 | ||
FQ229524.1 | - | 1 - 2 | 67 | 151 | ||
U07971.1 | - | 8 - 9 | 1212 | 151 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
