Assay Details
Gene Symbol: Slc2a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031741.1 | NP_113929.1 | 9 - 10 | 1137 | 86 | |
GenBank mRNA | BC076378.1 | - | 9 - 10 | 1122 | 86 | |
BC161843.1 | - | 9 - 10 | 1124 | 86 | ||
D13871.1 | - | 9 - 10 | 1137 | 86 | ||
D28562.1 | - | 9 - 10 | 1096 | 86 | ||
L05195.1 | - | 9 - 10 | 1109 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
