Assay Details
Gene Symbol: Casp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271984.1 | NP_001258913.1 | 5 - 6 | 492 | 95 | |
NM_031775.3 | NP_113963.2 | 6 - 7 | 647 | 95 | ||
XM_008761527.1 | XP_008759749.1 | 4 - 5 | 437 | 95 | ||
XM_008761528.1 | XP_008759750.1 | 4 - 5 | 437 | 95 | ||
GenBank mRNA | AF025670.1 | - | 6 - 7 | 665 | 95 | |
BC078785.1 | - | 6 - 7 | 612 | 95 | ||
DY310578.1 | - | 5 - 6 | 479 | 95 | ||
FM068086.1 | - | 5 - 6 | 492 | 95 |
Gene Symbol: LOC103689977 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006233278.2 | XP_006233340.1 | 6 - 7 | 672 | 95 | |
XM_006233279.2 | XP_006233341.1 | 5 - 6 | 494 | 95 | ||
XM_008761476.1 | XP_008759698.1 | 6 - 7 | 674 | 95 | ||
XM_008761477.1 | XP_008759699.1 | 5 - 6 | 497 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
