Assay Details
Gene Symbol: Ppig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031793.1 | NP_113981.1 | 13 - 14 | 1352 | 119 | |
XM_006234324.2 | XP_006234386.1 | 13 - 14 | 1728 | 119 | ||
XM_006234325.2 | XP_006234387.1 | 13 - 14 | 1549 | 119 | ||
XM_006234326.2 | XP_006234388.1 | 13 - 14 | 1672 | 119 | ||
XM_006234327.2 | XP_006234389.1 | 12 - 13 | 1624 | 119 | ||
XM_006234328.2 | XP_006234390.1 | 13 - 14 | 1619 | 119 | ||
XM_006234330.2 | XP_006234392.1 | 12 - 13 | 1445 | 119 | ||
XM_006234331.2 | XP_006234393.1 | 12 - 13 | 1571 | 119 | ||
XM_006234333.2 | XP_006234395.1 | 12 - 13 | 1585 | 119 | ||
XM_006234334.2 | XP_006234396.1 | 12 - 13 | 1406 | 119 | ||
XM_006234335.2 | XP_006234397.1 | 12 - 13 | 1532 | 119 | ||
XM_008761930.1 | XP_008760152.1 | 12 - 13 | 1648 | 119 | ||
XM_008761931.1 | XP_008760153.1 | 12 - 13 | 1609 | 119 | ||
GenBank mRNA | AF043642.1 | - | 13 - 14 | 1352 | 119 | |
FQ230786.1 | - | 8 - 9 | 867 | 119 | ||
FQ232477.1 | - | 8 - 9 | 868 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
