Assay Details
Gene Symbol: Ppig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031793.1 | NP_113981.1 | 1 - 2 | 147 | 87 | |
GenBank mRNA | AF043642.1 | - | 1 - 2 | 147 | 87 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF043642.1, NM_031793.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
