Assay Details
Gene Symbol: Asah1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053407.3 | NP_445859.2 | 11 - 12 | 955 | 113 | |
GenBank mRNA | AF214647.1 | - | 11 - 12 | 935 | 113 | |
BC061540.1 | - | 11 - 12 | 958 | 6919949 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
