Assay Details
Gene Symbol: Tinagl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053582.2 | NP_446034.1 | 8 - 9 | 1099 | 84 | |
XM_006238977.2 | XP_006239039.1 | 9 - 10 | 84 | |||
XM_006238978.2 | XP_006239040.1 | 9 - 10 | 84 | |||
GenBank mRNA | AB050717.1 | - | 8 - 9 | 1100 | 84 | |
BC097299.1 | - | 9 - 10 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
