Assay Details
Gene Symbol: Pip4k2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053926.2 | NP_446378.1 | 3 - 4 | 434 | 59 | |
GenBank mRNA | AB032899.1 | - | 3 - 4 | 434 | 59 |
Gene Symbol: LOC100909950 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006254325.2 | XP_006254387.1 | 2 - 3 | 449 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
