Assay Details
Gene Symbol: Lef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130429.1 | NP_569113.1 | 4 - 5 | 540 | 77 | |
XM_006233302.2 | XP_006233364.1 | 4 - 5 | 77 | |||
XM_006233303.2 | XP_006233365.1 | 4 - 5 | 77 | |||
XM_006233305.2 | XP_006233367.1 | 4 - 5 | 77 | |||
XM_006233306.2 | XP_006233368.1 | 4 - 5 | 77 | |||
XM_006233309.2 | XP_006233371.1 | 4 - 5 | 77 | |||
XM_008761489.1 | XP_008759711.1 | 4 - 5 | 77 | |||
XM_008761490.1 | XP_008759712.1 | 4 - 5 | 77 | |||
XM_008761491.1 | XP_008759713.1 | 4 - 5 | 77 | |||
XM_008761492.1 | XP_008759714.1 | 4 - 5 | 77 | |||
GenBank mRNA | AF198533.1 | - | 4 - 5 | 540 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
