Assay Details
Gene Symbol: Dkc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133419.1 | NP_596910.1 | 6 - 7 | 603 | 149 | |
XM_008773596.1 | XP_008771818.1 | 6 - 7 | 1049 | 149 | ||
GenBank mRNA | BC087600.1 | - | 6 - 7 | 562 | 149 | |
BC099832.1 | - | 6 - 7 | 561 | 149 | ||
Z34922.1 | - | 6 - 7 | 603 | 149 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
