Assay Details
Gene Symbol: Il13ra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133538.2 | NP_598222.2 | 13 - 14 | 1715 | 162 | |
XM_006257427.2 | XP_006257489.1 | 12 - 13 | 1583 | 162 | ||
XM_006257428.2 | XP_006257490.1 | 12 - 13 | 1701 | 162 | ||
XM_006257430.2 | XP_006257492.1 | 11 - 12 | 1514 | 162 | ||
XM_006257431.2 | XP_006257493.1 | 9 - 10 | 1222 | 162 | ||
XM_008773388.1 | XP_008771610.1 | 12 - 13 | 1855 | 162 | ||
XM_008773389.1 | XP_008771611.1 | 13 - 14 | 1742 | 162 | ||
XM_008773390.1 | XP_008771612.1 | 9 - 10 | 1316 | 162 | ||
XM_008773391.1 | XP_008771613.1 | 9 - 10 | 1923 | 162 | ||
GenBank mRNA | AF448818.1 | - | 13 - 14 | 1715 | 162 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
