Assay Details
Gene Symbol: Rabggtb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138708.2 | NP_619715.1 | 2 - 3 | 315 | 115 | |
GenBank mRNA | BC060536.1 | - | 339 | 115 | ||
BC078683.1 | - | 2 - 3 | 304 | 115 | ||
FQ214773.1 | - | 319 | 115 | |||
FQ216744.1 | - | 319 | 115 | |||
FQ224712.1 | - | 318 | 115 | |||
FQ229109.1 | - | 319 | 115 | |||
L10416.1 | - | 2 - 3 | 315 | 115 | ||
S62097.1 | - | 2 - 3 | 315 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
