Assay Details
Gene Symbol: Snrk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138833.1 | NP_620188.1 | 1 - 2 | 43 | 154 | |
XM_006244057.2 | XP_006244119.1 | 2 - 3 | 89 | 154 | ||
XM_006244058.2 | XP_006244120.1 | 2 - 3 | 123 | 154 | ||
XM_006244059.1 | XP_006244121.1 | 2 - 3 | 82 | 154 | ||
GenBank mRNA | X89383.1 | - | 1 - 2 | 43 | 154 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_138833.1, X89383.1, XM_006244057.2, XM_006244058.2, XM_006244059.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
