Assay Details
Gene Symbol: Bmf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139258.1 | NP_640351.1 | 3 - 4 | 537 | 66 | |
XM_006234723.2 | XP_006234785.1 | 3 - 4 | 575 | 66 | ||
XM_006234724.2 | XP_006234786.1 | 3 - 4 | 578 | 66 | ||
GenBank mRNA | AF506761.1 | - | 3 - 4 | 537 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
