Assay Details
Gene Symbol: Arfgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145090.3 | NP_659558.1 | 3 - 4 | 344 | 98 | |
XM_006235731.2 | XP_006235793.1 | 3 - 4 | 444 | 98 | ||
XM_006235732.1 | XP_006235794.1 | 3 - 4 | 385 | 98 | ||
XM_006235733.2 | XP_006235795.1 | 2 - 3 | 653 | 98 | ||
XM_006235734.2 | XP_006235796.1 | 2 - 3 | 652 | 98 | ||
GenBank mRNA | BC070895.1 | - | 3 - 4 | 325 | 7099840 | 98 |
DQ167810.1 | - | 1 - 2 | 61 | 98 | ||
DQ167811.1 | - | 1 - 2 | 61 | 98 | ||
FQ222295.1 | - | 3 - 4 | 367 | 98 | ||
FQ232032.1 | - | 3 - 4 | 365 | 98 | ||
FQ233265.1 | - | 3 - 4 | 365 | 98 | ||
U35776.1 | - | 3 - 4 | 344 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -



Gene Ontology Categories:
