Assay Details
Gene Symbol: Arfgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145090.3 | NP_659558.1 | 8 - 9 | 911 | 57 | |
XM_006235731.2 | XP_006235793.1 | 8 - 9 | 1011 | 57 | ||
XM_006235732.1 | XP_006235794.1 | 8 - 9 | 952 | 57 | ||
XM_006235733.2 | XP_006235795.1 | 7 - 8 | 1220 | 57 | ||
XM_006235734.2 | XP_006235796.1 | 7 - 8 | 1219 | 57 | ||
GenBank mRNA | BC070895.1 | - | 8 - 9 | 892 | 7099840 | 57 |
DQ167810.1 | - | 6 - 7 | 628 | 57 | ||
DQ167811.1 | - | 6 - 7 | 628 | 57 | ||
FQ222295.1 | - | 8 - 9 | 934 | 57 | ||
FQ232032.1 | - | 8 - 9 | 932 | 57 | ||
FQ233265.1 | - | 8 - 9 | 932 | 57 | ||
U35776.1 | - | 8 - 9 | 911 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -



Gene Ontology Categories:
