Assay Details
Gene Symbol: Aurka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153296.2 | NP_695208.2 | 1 - 2 | 47 | 91 | |
XM_006235675.2 | XP_006235737.1 | 2 - 3 | 126 | 91 | ||
XM_006235676.2 | XP_006235738.1 | 2 - 3 | 143 | 91 | ||
GenBank mRNA | AF537333.1 | - | 1 - 2 | 35 | 91 | |
BC104676.1 | - | 1 - 2 | 47 | 7453115 | 91 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
