Assay Details
Gene Symbol: Pafah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177932.2 | NP_808793.1 | 1 - 2 | 98 | 85 | |
XM_006239127.2 | XP_006239189.1 | 2 - 3 | 399 | 85 | ||
XM_008764144.1 | XP_008762366.1 | 2 - 3 | 401 | 85 | ||
GenBank mRNA | AY225592.1 | - | 1 - 2 | 98 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
