Assay Details
Gene Symbol: Pafah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177932.2 | NP_808793.1 | 2 - 3 | 257 | 93 | |
XM_006239127.2 | XP_006239189.1 | 3 - 4 | 558 | 93 | ||
XM_008764144.1 | XP_008762366.1 | 3 - 4 | 560 | 93 | ||
GenBank mRNA | AY225592.1 | - | 2 - 3 | 257 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
