Assay Details
Gene Symbol: Myrip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182844.1 | NP_878264.1 | 1 - 2 | 105 | 84 | |
XM_006244110.2 | XP_006244172.1 | 1 - 2 | 145 | 84 | ||
XM_008766684.1 | XP_008764906.1 | 1 - 2 | 145 | 84 | ||
XM_008766685.1 | XP_008764907.1 | 2 - 3 | 259 | 84 | ||
GenBank mRNA | AY331984.1 | - | 1 - 2 | 105 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
