Assay Details
Gene Symbol: Stk40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183056.2 | NP_898879.2 | 1 - 2 | 149 | 56 | |
GenBank mRNA | AY336055.1 | - | 1 - 2 | 151 | 56 | |
BC100062.1 | - | 1 - 2 | 149 | 7124923 | 56 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AY336055.1,BC100062.1,NM_183056.2 transcripts.
This assay is in the 5'UTR region of the AY336055.1,BC100062.1,NM_183056.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
