Assay Details
Gene Symbol: Stk40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183056.2 | NP_898879.2 | 9 - 10 | 1167 | 156 | |
XM_006238881.2 | XP_006238943.1 | 9 - 10 | 2877 | 156 | ||
GenBank mRNA | AY336055.1 | - | 9 - 10 | 1169 | 156 | |
BC100062.1 | - | 9 - 10 | 1167 | 7124923 | 156 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
