Assay Details
Gene Symbol: Tpd52l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198744.2 | NP_942039.1 | 5 - 6 | 627 | 86 | |
XM_006235760.2 | XP_006235822.1 | 4 - 5 | 581 | 86 | ||
GenBank mRNA | BC059167.1 | - | 5 - 6 | 628 | 6890828 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
