Assay Details
Gene Symbol: Fgfrl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199114.1 | NP_954545.1 | 1 - 2 | 215 | 133 | |
GenBank mRNA | AJ536020.1 | - | 1 - 2 | 215 | 133 | |
BC097261.1 | - | 1 - 2 | 171 | 133 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
