Assay Details
Gene Symbol: Clca4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201419.1 | NP_958822.1 | 2 - 3 | 219 | 76 | |
GenBank mRNA | AY333961.1 | - | 2 - 3 | 219 | 76 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AY333961.1, NM_201419.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
