Assay Details
Gene Symbol: Ptpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012637.2 | NP_036769.1 | 6 - 7 | 774 | 62 | |
XM_006235638.2 | XP_006235700.1 | 6 - 7 | 843 | 62 | ||
XM_006235639.2 | XP_006235701.1 | 6 - 7 | 844 | 62 | ||
GenBank mRNA | BC081829.1 | - | 6 - 7 | 774 | 7135855 | 62 |
M33962.1 | - | 6 - 7 | 816 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
