Assay Details
Gene Symbol: Spp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012881.2 | NP_037013.2 | 2 - 3 | 161 | 88 | |
XM_008769996.1 | XP_008768218.1 | 2 - 3 | 234 | 88 | ||
GenBank mRNA | BC078874.1 | - | 2 - 3 | 141 | 7110677 | 88 |
CV102664.1 | - | 2 - 3 | 161 | 88 | ||
FQ213473.1 | - | 2 - 3 | 150 | 88 | ||
FQ222192.1 | - | 2 - 3 | 177 | 88 | ||
M14656.1 | - | 2 - 3 | 134 | 88 | ||
M99252.1 | - | 2 - 3 | 170 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC078874.1, M14656.1, M99252.1, NM_012881.2, XM_008769996.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
