Assay Details
Gene Symbol: Gfra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012959.1 | NP_037091.1 | 6 - 7 | 1135 | 100 | |
XM_008760512.1 | XP_008758734.1 | 7 - 8 | 1278 | 100 | ||
XM_008760513.1 | XP_008758735.1 | 7 - 8 | 1142 | 100 | ||
XM_008760514.1 | XP_008758736.1 | 5 - 6 | 1158 | 100 | ||
GenBank mRNA | AJ002072.1 | - | 5 - 6 | 864 | 100 | |
U59486.1 | - | 6 - 7 | 1180 | 100 | ||
U97142.1 | - | 6 - 7 | 1135 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
