Assay Details
Gene Symbol: Pcmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013073.3 | NP_037205.2 | 2 - 3 | 264 | 107 | |
GenBank mRNA | BC088417.1 | - | 2 - 3 | 264 | 107 | |
D11475.1 | - | 2 - 3 | 124 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
