Assay Details
Gene Symbol: Tnfrsf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013091.1 | NP_037223.1 | 2 - 3 | 429 | 96 | |
GenBank mRNA | AF329976.1 | - | 2 - 3 | 428 | 96 | |
AF329977.1 | - | 2 - 3 | 428 | 96 | ||
AF329978.1 | - | 2 - 3 | 428 | 96 | ||
AF329979.1 | - | 2 - 3 | 428 | 96 | ||
AF329980.1 | - | 2 - 3 | 428 | 96 | ||
AF329981.1 | - | 2 - 3 | 428 | 96 | ||
BC086413.1 | - | 2 - 3 | 449 | 7110094 | 96 | |
M63122.1 | - | 2 - 3 | 429 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
