Assay Details
Gene Symbol: Dnase1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013097.2 | NP_037229.1 | 4 - 5 | 598 | 92 | |
XM_006245829.2 | XP_006245891.1 | 5 - 6 | 1278 | 92 | ||
GenBank mRNA | AF397150.1 | - | 4 - 5 | 598 | 92 | |
AF397151.1 | - | 5 - 6 | 469 | 92 | ||
BC089764.1 | - | 4 - 5 | 598 | 7130586 | 92 | |
X56060.1 | - | 4 - 5 | 451 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
