Assay Details
Gene Symbol: Slc19a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017299.2 | NP_058995.2 | 1 - 2 | 201 | 153 | |
GenBank mRNA | AF099009.1 | - | 1 - 2 | 210 | 153 | |
AF099010.1 | - | 1 - 2 | 210 | 153 | ||
U38180.1 | - | 1 - 2 | 194 | 146 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF099009.1, AF099010.1, NM_017299.2, U38180.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
