Assay Details
Gene Symbol: Coq3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019187.1 | NP_062060.1 | 1 - 2 | 67 | 103 | |
GenBank mRNA | BC081811.1 | - | 232 | 103 | ||
BC098666.1 | - | 225 | 103 | |||
BC126094.1 | - | 239 | 103 | |||
FQ216025.1 | - | 279 | 103 | |||
L20427.1 | - | 1 - 2 | 67 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
