Assay Details
Gene Symbol: Mapk13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019231.2 | NP_062104.2 | 11 - 12 | 1083 | 76 | |
GenBank mRNA | AF092534.1 | - | 11 - 12 | 1025 | 76 | |
BC107849.1 | - | 11 - 12 | 1083 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
