Assay Details
Gene Symbol: Fgb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020071.2 | NP_064456.2 | 2 - 3 | 311 | 67 | |
XM_006232524.2 | XP_006232586.1 | 2 - 3 | 329 | 67 | ||
XM_008761078.1 | XP_008759300.1 | 2 - 3 | 268 | 67 | ||
GenBank mRNA | AY321323.1 | - | 307 | 67 | ||
AY325147.1 | - | 307 | 67 | |||
AY325153.1 | - | 307 | 67 | |||
BC088102.1 | - | 2 - 3 | 311 | 7371665 | 67 | |
FQ209924.1 | - | 2 - 3 | 318 | 67 | ||
FQ219024.1 | - | 2 - 3 | 221 | 67 | ||
U05675.1 | - | 2 - 3 | 312 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
