Assay Details
Gene Symbol: Smad5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021692.1 | NP_067724.1 | 3 - 4 | 854 | 87 | |
XM_006253586.2 | XP_006253648.1 | 6 - 7 | 1319 | 87 | ||
XM_006253587.2 | XP_006253649.1 | 5 - 6 | 1237 | 87 | ||
XM_006253588.2 | XP_006253650.1 | 6 - 7 | 1561 | 87 | ||
XM_008771446.1 | XP_008769668.1 | 5 - 6 | 2897 | 87 | ||
XM_008771447.1 | XP_008769669.1 | 6 - 7 | 2079 | 87 | ||
XM_008771448.1 | XP_008769670.1 | 6 - 7 | 2254 | 87 | ||
GenBank mRNA | AB010955.1 | - | 3 - 4 | 854 | 87 | |
BC161849.1 | - | 4 - 5 | 1041 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
