Assay Details
Gene Symbol: Htr3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022189.1 | NP_071525.1 | 6 - 7 | 720 | 65 | |
GenBank mRNA | AF155044.1 | - | 6 - 7 | 720 | 65 | |
AF303447.1 | - | 387 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
