Assay Details
Gene Symbol: Htr3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022189.1 | NP_071525.1 | 7 - 8 | 922 | 72 | |
GenBank mRNA | AF155044.1 | - | 7 - 8 | 922 | 72 | |
AF303447.1 | - | 589 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
serotonin receptor signaling pathway
ion transmembrane transport
cellular response to growth factor stimulus
cation transmembrane transport
G protein-coupled serotonin receptor activity
extracellular ligand-gated ion channel activity
serotonin-gated cation-selective channel activity
serotonin receptor signaling pathway
ion transmembrane transport
cellular response to growth factor stimulus
cation transmembrane transport
G protein-coupled serotonin receptor activity
extracellular ligand-gated ion channel activity
serotonin-gated cation-selective channel activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P014080 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934027 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432338 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439903 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238357 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451959 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330283 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605204 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410206 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861677 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621132 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881697 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337982 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716434 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151293 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116543 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89615 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P014080 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P014080 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P014080 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P478850 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P12153 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 64918 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 18804 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 68751 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 20613 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10917137 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10917138 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10917143 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10917139 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10917140 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10917141 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10917136 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10917142 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10917144 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10917135 | - | - |
Rat Genome 230 2.0 Array | 1387602_a_at | - | - |
Rat Genome 230 2.0 Array | 1371077_PM_at | - | - |
Rat Genome 230 2.0 Array | 1371077_at | - | - |
Rat Genome 230 2.0 Array | 1387602_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1371077_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1387602_a_at | - | - |
Affymetrix HT Rat Focus Array | 1387602_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1371077_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0800020003.rn.1 | TC0800002491.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0800020002.rn.1 | TC0800002491.rn.1 | RTA TC |
Rat Expression Set 230 | 1387602_a_at | - | - |
Rat Expression Set 230 | 1371077_PM_at | - | - |
Rat Expression Set 230 | 1387602_PM_a_at | - | - |
Rat Expression Set 230 | 1371077_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10917135 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21934142 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1356352 | - | - |
RatRef-12 Expression BeadChip | ILMN_48155 | - | - |
