Assay Details
Gene Symbol: Clcn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022198.2 | NP_071534.2 | 4 - 5 | 500 | 159 | |
XM_006256828.2 | XP_006256890.1 | 4 - 5 | 803 | 159 | ||
GenBank mRNA | BC092209.1 | - | 4 - 5 | 500 | 7315063 | 159 |
FQ210965.1 | - | 4 - 5 | 539 | 159 | ||
Z36944.1 | - | 2 - 3 | 208 | 159 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
