Assay Details
Gene Symbol: Hpse | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022605.1 | NP_072127.1 | 2 - 3 | 417 | 78 | |
GenBank mRNA | AF184967.1 | - | 2 - 3 | 417 | 78 | |
AF359508.1 | - | 2 - 3 | 444 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
