Assay Details
Gene Symbol: Msn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030863.1 | NP_110490.1 | 8 - 9 | 1057 | 99 | |
GenBank mRNA | AF004811.1 | - | 8 - 9 | 1057 | 99 | |
FQ230914.1 | - | 1144 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
