Assay Details
Gene Symbol: Gtf2f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031042.3 | NP_112304.2 | 2 - 3 | 251 | 118 | |
GenBank mRNA | BC091112.1 | - | 2 - 3 | 251 | 118 | |
D10665.1 | - | 2 - 3 | 301 | 118 | ||
FQ232071.1 | - | 2 - 3 | 290 | 118 | ||
FQ233324.1 | - | 2 - 3 | 290 | 118 | ||
L01267.1 | - | 2 - 3 | 228 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
