Assay Details
Gene Symbol: Apba3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031781.1 | NP_113969.1 | 3 - 4 | 832 | 86 | |
XM_006241018.2 | XP_006241080.1 | 4 - 5 | 993 | 86 | ||
XM_006241019.2 | XP_006241081.1 | 4 - 5 | 966 | 86 | ||
XM_006241020.2 | XP_006241082.1 | 4 - 5 | 991 | 86 | ||
GenBank mRNA | AF029109.1 | - | 3 - 4 | 832 | 86 | |
BC161842.1 | - | 4 - 5 | 894 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
