Assay Details
Gene Symbol: Slc1a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032065.1 | NP_114454.1 | 7 - 8 | 1504 | 82 | |
XM_008765182.1 | XP_008763404.1 | 7 - 8 | 1678 | 82 | ||
GenBank mRNA | AY286330.1 | - | 7 - 8 | 1498 | 82 | |
BC098912.1 | - | 7 - 8 | 1528 | 82 | ||
U89608.1 | - | 7 - 8 | 1504 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
