Assay Details
Gene Symbol: Ralbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032067.1 | NP_114456.1 | 64 | 74 | ||
GenBank mRNA | BC089788.1 | - | 118 | 74 | ||
U82623.1 | - | 64 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC089788.1, NM_032067.1, U82623.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
