Assay Details
Gene Symbol: Psmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033236.1 | NP_150239.1 | 2 - 3 | 128 | 146 | |
GenBank mRNA | BC061542.1 | - | 2 - 3 | 170 | 146 | |
D50694.1 | - | 2 - 3 | 128 | 146 | ||
FQ212821.1 | - | 2 - 3 | 158 | 146 | ||
FQ230973.1 | - | 2 - 3 | 187 | 146 | ||
U13895.1 | - | 2 - 3 | 73 | 146 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
