Assay Details
Gene Symbol: Chrna3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052805.2 | NP_434692.2 | 2 - 3 | 205 | 73 | |
GenBank mRNA | AY574254.1 | - | 2 - 3 | 205 | 73 | |
L31621.1 | - | 2 - 3 | 281 | 73 | ||
X03440.1 | - | 2 - 3 | 355 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
