Assay Details
Gene Symbol: Gne | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053765.2 | NP_446217.1 | 7 - 8 | 1616 | 141 | |
XM_006238006.2 | XP_006238068.1 | 7 - 8 | 1434 | 141 | ||
XM_006238008.1 | XP_006238070.1 | 7 - 8 | 1400 | 141 | ||
XM_006238009.2 | XP_006238071.1 | 7 - 8 | 1777 | 141 | ||
GenBank mRNA | BC062011.1 | - | 7 - 8 | 1616 | 5621384 | 141 |
FQ224587.1 | - | 6 - 7 | 1279 | 141 | ||
Y07744.1 | - | 6 - 7 | 1324 | 141 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
