Assay Details
Gene Symbol: Lpar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF090347.1 | - | 1 - 2 | 55 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF090347.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
